4-Iodo-4'-nitrobiphenyl ≥98.0%
Supplier: TCI America
I0537-1G
I0537-5G
TCI0537-001GEA
45.82
CAD
TCI0537-001G
TCI0537-005G
4-Iodo-4'-nitrobiphenyl ≥98.0%
4-Iodo-4'-nitrobiphenyl
CAS Number: 29170-08-9
MDL Number: MFCD00024698
Molecular Formula: C12H8INO2
Molecular Weight: 325.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 216
Formula:
C₁₂H₈INO₂ Melting Pt: 216 °C |
MDL Number:
MFCD00024698 CAS Number: 29170-08-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...