Indocyanine green

Supplier: TCI America

Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate

I0535-100MG I0535-1G
TCI0535-100MGEA 302.1 CAD
TCI0535-100MG TCI0535-001G
Indocyanine green
Indocyanine green

CAS Number: 3599-32-4
MDL Number: MFCD00013078
Molecular Formula: C43H48N2O6S2
Molecular Weight: 774.97
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Cardio-green, Foxgreen, ICG

Formula: C₄₃H₄₇N₂NaO₆S₂
MDL Number: MFCD00013078
CAS Number: 3599-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20193a6b42b82fd9c20e174e3a4f29ec7e6646c3&vid=20193a6b42b82fd9c20e174e3a4f29ec7e6646c3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR