2-Iodopyridine ≥96.0% stabilized
Supplier: TCI America
I0533-5G
I0533-25G
TCI0533-005GEA
104.28
CAD
TCI0533-005G
TCI0533-025G
2-Iodopyridine ≥96.0% stabilized
2-Iodopyridine
(stabilized with Na2S2O3)
CAS Number: 5029-67-4
MDL Number: MFCD00464928
Molecular Formula: C5H4IN
Molecular Weight: 205.00
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Specific Gravity (20/20): 1.98
Formula:
C₅H₄IN Boiling Pt: 102 °C Melting Pt: 118…120 °C Density: 1.98 (20 °C) |
MDL Number:
MFCD00464928 CAS Number: 5029-67-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...