2-Indancarboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydro-1H-indene-2-carboxylic acid, Indan-2-carboxylic acid

I0532-1G I0532-5G
TCI0532-001GEA 49.61 CAD
TCI0532-001G TCI0532-5G
2-Indancarboxylic acid ≥98.0% (by titrimetric analysis)
2-Indancarboxylic acid

CAS Number: 25177-85-9
MDL Number: MFCD00085095
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 128

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 192 °C
Melting Pt: 128 °C
MDL Number: MFCD00085095
CAS Number: 25177-85-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b007217f701ef2cecb5cbae8e316bbdf675bb6c&vid=3b007217f701ef2cecb5cbae8e316bbdf675bb6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR