Methyl-2-amino-5-iodobenzoate ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-5-iodobenzoic acid methyll ester, Methyl 2-amino-5-iodobenzoate, Methyl 5-Iodoanthranilate, Methyl 2-Amino-5-iodobenzoate, 5-Iodoanthranilic acid methyll ester

I0516-25G I0516-5G
TCI0516-25GEA 219.62 CAD
TCI0516-25G TCI0516-005G
Methyl-2-amino-5-iodobenzoate ≥98.0%
Methyl 2-amino-5-iodobenzoate

CAS Number: 77317-55-6
MDL Number: MFCD00798574
Molecular Formula: C8H8INO2
Molecular Weight: 277.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 2-Amino-5-iodobenzoic Acid Methyl Ester, 5-Iodoanthranilic Acid Methyl Ester, Methyl 2-Amino-5-iodobenzoate

Formula: C₈H₈INO₂
MW: 277.0591 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00798574
CAS Number: 77317-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38d82ef012e8392dc285d70679c2793d3d49be9c&vid=38d82ef012e8392dc285d70679c2793d3d49be9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR