Isoquinoline-3-carboxylic acid monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0509-1G I0509-5G
TCI0509-001GEA 226.41 CAD
TCI0509-001G TCI0509-5G
Isoquinoline-3-carboxylic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Isoquinoline-3-carboxylic acid monohydrate

CAS Number: 203626-75-9
MDL Number: MFCD00075137
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 167

Formula: C₁₀H₉NO₃
Melting Pt: 167 °C
MDL Number: MFCD00075137
CAS Number: 207399-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9059005d0a556fceff7e325fa56f209cb9d0fac&vid=e9059005d0a556fceff7e325fa56f209cb9d0fac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR