Ethyl-cis-3-iodoacrylate ≥98.0% stabilized

Supplier: TCI America

I0502-5G I0502-25G
TCI0502-005GEA 314.42 CAD
TCI0502-005G TCI0502-25G
Ethyl-cis-3-iodoacrylate ≥98.0% stabilized
Ethyl-cis-3-iodoacrylate

(stabilized with Copper chip)
CAS Number: 31930-36-6
MDL Number: MFCD00209584
Molecular Formula: C5H7IO2
Molecular Weight: 226.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 99
Specific Gravity (20/20): 1.78
Storage Temperature: 0-10°C


Other Product Names / Synonyms: cis-3-Iodoacrylic Acid Ethyl Ester

Formula: C₅H₇IO₂
Boiling Pt: 85 °C
Density: 1.78 (20 °C)
Flash Pt: 99 °C
MDL Number: MFCD00209584
CAS Number: 31930-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1c81c925cc328e13fdbacd95b15431ce5293088&vid=a1c81c925cc328e13fdbacd95b15431ce5293088&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR