Isopropyl methyl sulfone ≥97.0%
Supplier: TCI America
I0484-25G
I0484-5G
TCI0484-025GEA
515.08
CAD
TCI0484-025G
TCI0484-5G
Isopropyl methyl sulfone ≥97.0%
Isopropyl methyl sulfone
CAS Number: 4853-74-1
MDL Number: MFCD00050415
Molecular Formula: C4H10O2S
Molecular Weight: 122.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.13
Formula:
C₄H₁₀O₂S Density: 1.13 (20 °C) |
MDL Number:
MFCD00050415 CAS Number: 4853-74-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...