(R)-2-Isopropylamino-2-phenylethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-2-(Isopropylamino)-2-phenylethanol

I0476-25G I0476-5G
TCI0476-25GEA 1118.64 CAD
TCI0476-25G TCI0476-005G
(R)-2-Isopropylamino-2-phenylethanol ≥98.0% (by GC, titration analysis)
(R)-2-Isopropylamino-2-phenylethanol

CAS Number: 112211-92-4
MDL Number: MFCD00671548
Molecular Formula: C11H17NO
Molecular Weight: 179.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 77

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00671548
CAS Number: 112211-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6000c7ea261ab90477f0f98644729c284a5fbd37&vid=941e18400d03d10442310ac8b3191bcbc86e54c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Specific rotation [a]20/D -65.0 to -73.0 deg (C=1,ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR