Methyl (S)-(-)-2-isocyanato-3-methylbutyrate ≥98.0%

Supplier: TCI America

Synonyms: Methyl (S)-(-)-2-Isocyanatoisovalerate, (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester

I0466-1G I0466-5G
TCI0466-001GEA 112.34 CAD
TCI0466-001G TCI0466-005G
Methyl (S)-(-)-2-isocyanato-3-methylbutyrate ≥98.0%
Methyl (S)-(-)-2-isocyanato-3-methylbutyrate

CAS Number: 30293-86-8
MDL Number: MFCD00191531
Molecular Formula: C7H11NO3
Molecular Weight: 157.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 78
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: -20 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
Boiling Pt: 76 °C
Density: 1.06 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00191531
CAS Number: 30293-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0219f52d3f12a55a4757ba18e08ba4fda3490e8&vid=a0219f52d3f12a55a4757ba18e08ba4fda3490e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR