L(+)-Isoleucinol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

I0462-5G I0462-1G
TCI0462-5GEA 372.88 CAD
TCI0462-5G TCI0462-001G
L(+)-Isoleucinol ≥97.0% (by GC, titration analysis)
L(+)-Isoleucinol

CAS Number: 24629-25-2
MDL Number: MFCD00004731
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 97
Melting point (°C): 42
Flash Point (°C): 100
Specific rotation [a]20/D: 2.5 deg (C=1.6, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,3S)-(+)-2-Amino-3-methyl-1-pentanol

Formula: C₆H₁₅NO
Boiling Pt: 97 °C
Melting Pt: 42 °C
Flash Pt: 100 °C
MDL Number: MFCD00004731
CAS Number: 24629-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=993155f09ce1e345d9f0cadadfcb9b2e77a8b4d1&vid=993155f09ce1e345d9f0cadadfcb9b2e77a8b4d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR