4,5-Bis(hydroxymethyl)imidazole ≥98.0%
Supplier: TCI America
Synonyms:
1H-Imidazole-4,5-dimethanol
I0457-1G
I0457-5G
TCI0457-001GEA
345.56
CAD
TCI0457-001G
TCI0457-5G
4,5-Bis(hydroxymethyl)imidazole ≥98.0%
4,5-Bis(hydroxymethyl)imidazole
CAS Number: 33457-48-6
MDL Number: MFCD00191661
Molecular Formula: C5H8N2O2
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 162
Other Product Names / Synonyms: 1H-Imidazole-4,5-dimethanol
Formula:
C₅H₈N₂O₂ MW: 128.13 g/mol Melting Pt: 162 °C |
MDL Number:
MFCD00191661 CAS Number: 33457-48-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...