Dimethyl-4,5-imidazoledicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl 1H-Imidazole-4,5-dicarboxylate, Dimethyl 4,5-imidazoledicarboxylate, 1H-Imidazole-4,5-dicarboxylic acid dimethyl ester

I0456-25G I0456-10G
TCI0456-25GEA 395 CAD
TCI0456-25G TCI0456-010G
Dimethyl-4,5-imidazoledicarboxylate ≥98.0%
Dimethyl-4,5-imidazoledicarboxylate

CAS Number: 3304-70-9
MDL Number: MFCD00051338
Molecular Formula: C7H8N2O4
Molecular Weight: 184.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1H-Imidazole-4,5-dicarboxylic Acid Dimethyl Ester

Formula: C₇H₈N₂O₄
MW: 184.1500 g/mol
MDL Number: MFCD00051338
CAS Number: 3304-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f87048aacd9026fd31c8cdbabb79bce3192d494&vid=8f87048aacd9026fd31c8cdbabb79bce3192d494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR