Diethyl acetonylmalonate ≥95.0%

Supplier: TCI America

A1503-1G A1503-5G
TCA1503-001GEA 59.46 CAD
TCA1503-001G TCA1503-005G
Diethyl acetonylmalonate ≥95.0%
Diethyl acetonylmalonate

CAS Number: 23193-18-2
MDL Number: MFCD00177949
Molecular Formula: C10H16O5
Molecular Weight: 216.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid


Other Product Names / Synonyms: Acetonylmalonic Acid Diethyl Ester, Diethyl 2-Oxopropylmalonate, 2-Oxopropylmalonic Acid Diethyl Ester

Formula: C₁₀H₁₆O₅
MDL Number: MFCD00177949
CAS Number: 23193-18-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0088099080632d6b2d8b9351cc92f17b71f6fd58&vid=0088099080632d6b2d8b9351cc92f17b71f6fd58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR