1-(Isocyanomethyl)-1H-benzotriazole ≥97.0% (by HPLC)

Supplier: TCI America

I0455-5G I0455-1G
TCI0455-5GEA 1056.23 CAD
TCI0455-5G TCI0455-001G
1-(Isocyanomethyl)-1H-benzotriazole ≥97.0% (by HPLC)
1-(Isocyanomethyl)-1H-benzotriazole

CAS Number: 87022-42-2
MDL Number: MFCD00961633
Molecular Formula: C8H6N4
Molecular Weight: 158.16
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Isonitriles (Multi-component Reaction)
Other Product Names / Synonyms: (1H-Benzotriazol-1-yl)methyl Isocyanide, 1-(Isocyanidomethyl)-1H-benzotriazole

Formula: C₈H₆N₄
MDL Number: MFCD00961633
CAS Number: 87022-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac4af41cda470777992352da8167fee3fa13275c&vid=ac4af41cda470777992352da8167fee3fa13275c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR