Diallyl isophthalate ≥98.0%

Supplier: TCI America

Synonyms: Isophthalic Acid Diallyl Ester

I0450-500G I0450-25G
TCI0450-500GEA 120.71 CAD
TCI0450-500G TCI0450-025G
Diallyl isophthalate ≥98.0%
Diallyl isophthalate

CAS Number: 1087-21-4
MDL Number: MFCD00048174
Molecular Formula: C14H14O4
Molecular Weight: 246.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 163
Specific Gravity (20/20): 1.13
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Isophthalic Acid Diallyl Ester

Formula: C₁₄H₁₄O₄
Boiling Pt: 163 °C
Density: 1.13 (20 °C)
Flash Pt: 163 °C
MDL Number: MFCD00048174
CAS Number: 1087-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61cb3e6592e8fd880114065c914d58634148a6e4&vid=61cb3e6592e8fd880114065c914d58634148a6e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR