4-Isobutylbenzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
I0437-10G
I0437-25G
TCI0437-010GEA
338.12
CAD
TCI0437-010G
TCI0437-25G
4-Isobutylbenzoic acid ≥99.0% (by GC, titration analysis)
4-Isobutylbenzoic acid
CAS Number: 38861-88-0
MDL Number: MFCD00191658
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Formula:
C₁₁H₁₄O₂ Melting Pt: 144 °C |
MDL Number:
MFCD00191658 CAS Number: 38861-88-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...