3-Isopropylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0433-25ML I0433-500ML
TCI0433-025MLEA 58.46 CAD
TCI0433-025ML TCI0433-500ML
3-Isopropylaniline ≥98.0% (by GC, titration analysis)
3-Isopropylaniline

CAS Number: 5369-16-4
MDL Number: MFCD00092918
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 102
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 3-Aminocumene, m-Cumidine

Formula: C₉H₁₃N
Boiling Pt: 80 °C
Density: 0.96 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00092918
CAS Number: 5369-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c54357facb655314dd02ef303337c7c0fc90730&vid=9c54357facb655314dd02ef303337c7c0fc90730&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR