4-Isopropyl-3-thiosemicarbazide
Supplier: TCI America
I0418-5G
I0418-1G
TCI0418-5GEA
261.17
CAD
TCI0418-5G
TCI0418-001G
4-Isopropyl-3-thiosemicarbazide
4-Isopropyl-3-thiosemicarbazide
CAS Number: 13431-36-2
MDL Number: MFCD00025144
Molecular Formula: C4H11N3S
Molecular Weight: 133.21
Form: Crystal
Color: White
Melting point (°C): 97
Formula:
C₄H₁₁N₃S Melting Pt: 97 °C |
MDL Number:
MFCD00025144 CAS Number: 13431-36-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...