Diphenyl isophthalate ≥99.0%

Supplier: TCI America

I0416-25G I0416-10G
TCI0416-25GEA 229.1 CAD
TCI0416-25G TCI0416-010G
Diphenyl isophthalate ≥99.0%
Diphenyl isophthalate

CAS Number: 744-45-6
MDL Number: MFCD00046051
Molecular Formula: C20H14O4
Molecular Weight: 318.33
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: Isophthalic Acid Diphenyl Ester

Formula: C₂₀H₁₄O₄
Melting Pt: 138 °C
MDL Number: MFCD00046051
CAS Number: 744-45-6

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR