Isomannide ≥98.0%

Supplier: TCI America

Synonyms: (3R,3aR,6R,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol, Isomannide, 1,4:3,6-Dianhydromannitol

I0406-25G I0406-5G
TCI0406-025GEA 152 CAD
TCI0406-025G TCI0406-5G
Isomannide ≥98.0%
Isomannide

CAS Number: 641-74-7
MDL Number: MFCD00064818
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 86
Specific rotation [a]20/D: 90 deg (C=3, H2O)


Product Categories: Bioscience, Sugars (Others)
Other Product Names / Synonyms: 1,4:3,6-Dianhydromannitol

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 150 °C
Melting Pt: 86 °C
MDL Number: MFCD00064818
CAS Number: 641-74-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e24900178a2cdbd95d6507ef2fa4a97f33e5e4&vid=a9e24900178a2cdbd95d6507ef2fa4a97f33e5e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR