Diethyl isopropylidenesuccinate ≥98.0%

Supplier: TCI America

I0404-1G I0404-5G
TCI0404-001GEA 132.72 CAD
TCI0404-001G TCI0404-005G
Diethyl isopropylidenesuccinate ≥98.0%
Diethyl isopropylidenesuccinate

CAS Number: 42103-98-0
MDL Number: MFCD00130132
Molecular Formula: C11H18O4
Molecular Weight: 214.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Isopropylidenesuccinic Acid Diethyl Ester

Formula: C₁₁H₁₈O₄
Boiling Pt: 103 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00130132
CAS Number: 42103-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=896f113a44cab7babb694930dfeb964fd46f2714&vid=896f113a44cab7babb694930dfeb964fd46f2714&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR