L-Alanyl-L-Proline

Supplier: TCI America

A1495-5G
TCA1495-005GEA 412.38 CAD
TCA1495-005G
L-Alanyl-L-Proline
Proteins and Peptides

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Ala-Pro-OH


  • Melting point: 171 °C
  • Specific rotation [a]20/D: –112 to –118° (C=2, H₂O)
  • Solubility in water: within almost transparency
  • EINECS: 236-795-8


Product Categories: Bioscience, Oligopeptides Other Product Names / Synonyms: H-Ala-Pro-OH

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b587a096155d7fceb2fe8febe2d0c45a3a078f09&vid=b587a096155d7fceb2fe8febe2d0c45a3a078f09&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR