p-Tolyl isobutyrate

Supplier: TCI America

I0396-25ML I0396-5ML
TCI0396-025MLEA 471.16 CAD
TCI0396-025ML TCI0396-5ML
p-Tolyl isobutyrate
p-Tolyl isobutyrate

CAS Number: 103-93-5
MDL Number: MFCD00026440
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 237
Flash Point (°C): 100
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Isobutyric Acid p-Tolyl Ester

Formula: C₁₁H₁₄O₂
Boiling Pt: 237 °C
Density: 1.00 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00026440
CAS Number: 103-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=335bccc61115eea48aa4281d2e54e953e8163362&vid=335bccc61115eea48aa4281d2e54e953e8163362&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR