(±)-Indoline-2-carboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: Indoline-2-carboxylic acid

I0394-25G I0394-5G
TCI0394-25GEA 230.68 CAD
TCI0394-25G TCI0394-5G
(±)-Indoline-2-carboxylic acid ≥97.0%
(±)-Indoline-2-carboxylic acid

CAS Number: 78348-24-0
MDL Number: MFCD00010635
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00010635
CAS Number: 78348-24-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a82e4e5a29d91787124dc6e177694b018b1c58d7&vid=a82e4e5a29d91787124dc6e177694b018b1c58d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR