O-Isobutylhydroxylamine hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
I0387-1G
I0387-5G
TCI0387-001GEA
197.5
CAD
TCI0387-001G
TCI0387-5G
O-Isobutylhydroxylamine hydrochloride ≥98.0% (by total nitrogen basis)
O-Isobutylhydroxylamine hydrochloride
CAS Number: 6084-58-8
MDL Number: MFCD00060208
Molecular Formula: C4H11NO
Molecular Weight: 125.60
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 129
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isobutyloxyamine Hydrochloride
Formula:
C₄H₁₂ClNO Melting Pt: 129 °C |
MDL Number:
MFCD00060208 CAS Number: 6084-58-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...