4,4'-Isopropylidenediphenoxyacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0356-5G I0356-25G
TCI0356-5GEA 330.22 CAD
TCI0356-5G TCI0356-025G
4,4'-Isopropylidenediphenoxyacetic acid ≥98.0% (by HPLC, titration analysis)
4,4'-Isopropylidenediphenoxyacetic acid

CAS Number: 3539-42-2
MDL Number: MFCD00060114
Molecular Formula: C19H20O6
Molecular Weight: 344.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 179


Product Categories: Materials Chemistry, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bisphenol A O,O-Diacetic Acid

Formula: C₁₉H₂₀O₆
Melting Pt: 179 °C
MDL Number: MFCD00060114
CAS Number: 3539-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481495b02370c466a553b39613f6c0cad44c9642&vid=481495b02370c466a553b39613f6c0cad44c9642&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR