Tris(2,3-dibromopropyl)isocyanurate ≥97.0%

Supplier: TCI America

Synonyms: 1,3,5-Tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione, Isocyanuric acid Tris(2,3-dibromopropyl) Ester

I0355-500G I0355-25G
TCI0355-500GEA 316 CAD
TCI0355-500G TCI0355-025G
Tris(2,3-dibromopropyl)isocyanurate ≥97.0%
Tris(2,3-dibromopropyl)isocyanurate

CAS Number: 52434-90-9
MDL Number: MFCD00010290
Molecular Formula: C12H15Br6N3O3
Molecular Weight: 728.69
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 110


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Isocyanuric Acid Tris(2,3-dibromopropyl) Ester

Formula: C₁₂H₁₅Br₆N₃O₃
MW: 728.69 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00010290
CAS Number: 52434-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c08be5de24925a9ebc95ec11ae5a0e0c8059f8aa&vid=c08be5de24925a9ebc95ec11ae5a0e0c8059f8aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR