Diethyl iminodiacetate ≥98.0%

Supplier: TCI America

Synonyms: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

I0326-25ML I0326-5ML
TCI0326-025MLEA 142.2 CAD
TCI0326-025ML TCI0326-5ML
Diethyl iminodiacetate ≥98.0%
Diethyl iminodiacetate

CAS Number: 6290-05-7
MDL Number: MFCD00041925
Molecular Formula: C8H15NO4
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Flash Point (°C): 113
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: Iminodiacetic Acid Diethyl Ester

Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Boiling Pt: 122 °C
Density: 1.08 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00041925
CAS Number: 6290-05-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37c5377f7edd44f0559dba9a1c304388f613867&vid=e37c5377f7edd44f0559dba9a1c304388f613867&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR