DL-Isoserine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-DL-IsoSer-OH, DL-3-Amino-2-hydroxypropionic Acid

I0309-100MG I0309-1G
TCI0309-100MGEA 30.02 CAD
TCI0309-100MG TCI0309-1G
DL-Isoserine ≥95.0% (by titrimetric analysis)
DL-Isoserine

CAS Number: 565-71-9
MDL Number: MFCD00008138
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 235


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-3-Amino-2-hydroxypropionic Acid

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00008138
CAS Number: 565-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4ca5e0e252494949d200b5bc8572aa51f5e743a&vid=a4ca5e0e252494949d200b5bc8572aa51f5e743a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR