(3-Acrylamidopropyl)trimethylammonium chloride ≥74-76% in water stabilised
Supplier: TCI America
Synonyms:
3-Acrylamido-N,N,N-trimethylpropan-1-aminium chloride
A1493-500ML
A1493-25ML
TCA1493-500MLEA
388.03
CAD
TCA1493-500ML
TCA1493-025ML
(3-Acrylamidopropyl)trimethylammonium chloride ≥74-76% in water stabilised
(3-Acrylamidopropyl)trimethylammonium chloride
(stabilized with MEHQ)
CAS Number: 45021-77-0
MDL Number: MFCD00274249
Molecular Formula: C9H19ClN2O
Molecular Weight: 206.71
Form: Clear Liquid
Color: Very Pale Yellow
Formula:
C₉H₁₉ClN₂O MW: 206.71 g/mol |
MDL Number:
MFCD00274249 CAS Number: 45021-77-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...