Iopanoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
I0300-5G
I0300-25G
TCI0300-5GEA
331.8
CAD
TCI0300-5G
TCI0300-025G
Iopanoic acid ≥98.0% (by titrimetric analysis)
Iopanoic acid
CAS Number: 96-83-3
MDL Number: MFCD00038687
Molecular Formula: C11H12I3NO2
Molecular Weight: 570.94
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: 3-Amino-alpha-ethyl-2,4,6-triiodohydrocinnamic Acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropionic Acid, Iodopanoic Acid, 2-(3-Amino-2,4,6-triiodobenzyl)butyric Acid
Formula:
C₁₁H₁₂I₃NO₂ Melting Pt: 153 °C |
MDL Number:
MFCD00038687 CAS Number: 96-83-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...