Monomethyl itaconate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate, 4-Methoxy-2-methylene-4-oxobutanoic acid
CAS Number: 7338-27-4
MDL Number: MFCD00021713
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 149
Melting point (°C): 72
Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Itaconic Acid Monomethyl Ester, beta-Methyl Itaconate, 4-Methyl Methylenesuccinate
Formula:
C₆H₈O₄ MW: 144.13 g/mol Boiling Pt: 149 °C Melting Pt: 72 °C |
MDL Number:
MFCD00021713 CAS Number: 7338-27-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...