Monobutyl itaconate ≥95.0%

Supplier: TCI America

Synonyms: 4-Butyl Methylenesuccinate, beta-Butyl Itaconate, Itaconic Acid Monobutyl Ester

I0267-25G I0267-500G
TCI0267-025GEA 57.84 CAD
TCI0267-025G TCI0267-500G
Monobutyl itaconate ≥95.0%
Monobutyl itaconate

CAS Number: 6439-57-2
MDL Number: MFCD00046533
Molecular Formula: C9H14O4
Molecular Weight: 186.21
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Freezing point (°C): 33


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: beta-Butyl Itaconate, 4-Butyl Methylenesuccinate, Itaconic Acid Monobutyl Ester

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
MDL Number: MFCD00046533
CAS Number: 6439-57-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86dd611c4c4821e1ee093fe6e7f8b9f727f5f16f&vid=86dd611c4c4821e1ee093fe6e7f8b9f727f5f16f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR