Acesulfame potassium ≥98.0%

Supplier: TCI America

Synonyms: 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide Potassium Salt, Potassium 6-methyl-4-oxo-4H-1,2,3-oxathiazin-3-ide 2,2-dioxide, Acesulphame K, Potassium 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide, Acesulfame K

A1490-25G A1490-500G A1490-100G
TCA1490-025GEA 108.45 CAD
TCA1490-025G TCA1490-500G TCA1490-100G
Acesulfame potassium ≥98.0%
Acesulfame potassium

CAS Number: 55589-62-3 MDL Number: MFCD00043833 Molecular Formula: C4H5NO4S Molecular Weight: 201.24 Purity/Analysis Method: 98.0% (T) Form: Crystal


Form: Crystal

Formula: C₄H₄KNO₄S
MW: 201.24 g/mol
MDL Number: MFCD00043833
CAS Number: 55589-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45b501048043d76894b86420c78b50fc4e9308ee&vid=45b501048043d76894b86420c78b50fc4e9308ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR