Isooctadecenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)

Supplier: TCI America

I0237-500G I0237-25G
TCI0237-500GEA 578.6 CAD
TCI0237-500G TCI0237-025G
Isooctadecenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)
Isooctadecenylsuccinic anhydride (mixture of branched chain isomers)

CAS Number: 58239-72-8
MDL Number: MFCD00070628
Molecular Formula: C22H38O3
Molecular Weight: 350.54
Purity/Analysis Method: >90.0% (T)
Form: Clear Liquid
Flash Point (°C): 201
Specific Gravity (20/20): 0.95

Formula: C₂₂H₃₈O₃
Density: 0.95 (20 °C)
Flash Pt: 201 °C
MDL Number: MFCD00070628
CAS Number: 58239-72-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed70ad798521e73cbf3cc3b89b79f4913000bcda&vid=ed70ad798521e73cbf3cc3b89b79f4913000bcda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR