Sodium isoascorbate monohydrate ≥98.0%

Supplier: TCI America

Synonyms: Isoascorbic acid Sodium salt, Sodium Erythorbate, Sodium (R)-2-((R)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Erythorbic acid Sodium salt, Sodium D-Araboascorbate, D-Araboascorbic acid Sodium salt

I0232-25G I0232-500G
TCI0232-025GEA 64.12 CAD
TCI0232-025G TCI0232-500G
Sodium isoascorbate monohydrate ≥98.0%
Sodium isoascorbate monohydrate

CAS Number: 6381-77-7
MDL Number: MFCD00067298
Molecular Formula: C6H8O6
Molecular Weight: 198.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 97 deg (C=10, H2O)


Product Categories: Bioscience, Vitamin Derivatives
Other Product Names / Synonyms: D-Araboascorbic Acid Sodium Salt, Erythorbic Acid Sodium Salt, Isoascorbic Acid Sodium Salt, Sodium D-Araboascorbate, Sodium Erythorbate

Formula: C₆H₈O₆
MW: 198.11 g/mol
MDL Number: MFCD00067298
CAS Number: 6381-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b655fb850cc4b3292a6757ce5b987c4f4625a679&vid=b655fb850cc4b3292a6757ce5b987c4f4625a679&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR