Isomaltose ≥97.0%

Supplier: TCI America

I0231-1G I0231-100MG
TCI0231-001GEA 543.3 CAD
TCI0231-001G TCI0231-100MG
Isomaltose ≥97.0%
Isomaltose

CAS Number: 499-40-1
MDL Number: MFCD00065373
Molecular Formula: C12H22O11
Molecular Weight: 342.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Specific rotation [a]20/D: 113 deg (C=0.7, H2O)


Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Disaccharides
Other Product Names / Synonyms: alpha-1,6-Glucobiose, 6-O-alpha-D-Glucopyranosyl-D-glucose

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00065373
CAS Number: 499-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14ca6941c053e5e99ff369562972ed5fbc6374b6&vid=14ca6941c053e5e99ff369562972ed5fbc6374b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR