2-(Isopropylamino)ethanol ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Isopropylethanolamine

I0227-25ML I0227-500ML I0227-100ML
TCI0227-025MLEA 99.22 CAD
TCI0227-025ML TCI0227-500ML TCI0227-100ML
2-(Isopropylamino)ethanol ≥99.0% (by GC, titration analysis)
2-(Isopropylamino)ethanol

CAS Number: 109-56-8
MDL Number: MFCD00041755
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 171
Flash Point (°C): 76
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: N-Isopropylethanolamine

Formula: C₅H₁₃NO
MW: 103.17 g/mol
Boiling Pt: 171 °C
Density: 0.90 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00041755
CAS Number: 109-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a479792ec7b540666ef849045bb5e1db9a2b538a&vid=a479792ec7b540666ef849045bb5e1db9a2b538a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR