Indigotrisulfonic acid potassium salt 55.0% (by HPLC)

Supplier: TCI America

I0220-1G I0220-5G
TCI0220-001GEA 88.4 CAD
TCI0220-001G TCI0220-005G
Indigotrisulfonic acid potassium salt 55.0% (by HPLC)
Indigotrisulfonic acid potassium salt

CAS Number: 67627-18-3
MDL Number: MFCD00013160
Molecular Formula: C16H10N2O11S3
Molecular Weight: 616.71
Purity/Analysis Method: >55.0% (HPLC)
Form: Crystal

Formula: C₁₆H₇K₃N₂O₁₁S₃
MDL Number: MFCD00013160
CAS Number: 67627-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=636014342f5eccfe57b635479c8f6b658623847b&vid=636014342f5eccfe57b635479c8f6b658623847b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 55.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR