Diethyl itaconate ≥98.0% stabilized

Supplier: TCI America

I0204-500G I0204-25G
TCI0204-500GEA 992.24 CAD
TCI0204-500G TCI0204-025G
Diethyl itaconate ≥98.0% stabilized
Diethyl itaconate

(stabilized with TBC)
CAS Number: 2409-52-1
MDL Number: MFCD00059230
Molecular Formula: C9H14O4
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 108
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Itaconic Acid Diethyl Ester

Formula: C₉H₁₄O₄
Boiling Pt: 228 °C
Density: 1.05 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00059230
CAS Number: 2409-52-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f11d15ef52d3853220afd1a9f411ce4ca6ff9520&vid=f11d15ef52d3853220afd1a9f411ce4ca6ff9520&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR