Dibutyl Itaconate ≥97.0% stabilized

Supplier: TCI America

I0201-500G I0201-25G
TCI0201-500GEA 115.34 CAD
TCI0201-500G TCI0201-025G
Dibutyl Itaconate ≥97.0% stabilized
Dibutyl Itaconate

(stabilized with HQ)
CAS Number: 2155-60-4
MDL Number: MFCD00027211
Molecular Formula: C13H22O4
Molecular Weight: 242.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 138
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: Itaconic Acid Dibutyl Ester

Formula: C₁₃H₂₂O₄
Boiling Pt: 190 °C
Density: 0.98 (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00027211
CAS Number: 2155-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8b2e6c7d870d8e9402c3bbf19f95a5461c280a3&vid=b8b2e6c7d870d8e9402c3bbf19f95a5461c280a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR