3-Methylbutyl isovalerate ≥98.0%
Supplier: TCI America
I0196-25ML
I0196-500ML
TCI0196-025MLEA
34.76
CAD
TCI0196-025ML
TCI0196-500ML
3-Methylbutyl isovalerate ≥98.0%
3-Methylbutyl isovalerate
CAS Number: 659-70-1
MDL Number: MFCD00048369
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 72
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: Isovaleric Acid Isoamyl Ester, Isopentyl Isovalerate, Isovaleric Acid Isopentyl Ester
Formula:
C₁₀H₂₀O₂ Boiling Pt: 194 °C Density: 0.86 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00048369 CAS Number: 659-70-1 EINECS: 211-536-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...