Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid) ≥90.0%

Supplier: TCI America

Synonyms: 2,2,4,8,10,10-Hexamethylundecane-5-carboxylic Acid, 2-(4,4-Dimethyl-2-pentyl)-5,7,7-trimethyl-n-octanoic Acid

I0184-500G I0184-25G
TCI0184-500GEA 315.34 CAD
TCI0184-500G TCI0184-025G
Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid) ≥90.0%
Isostearic acid (=2,2,4,8,10,10-Hexamethylundecane-5-carboxylic acid)

CAS Number: 54680-48-7
MDL Number: MFCD00135620
Molecular Formula: C18H36O2
Molecular Weight: 284.48
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 165
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: 2,2,4,8,10,10-Hexamethylundecane-5-carboxylic Acid, 2-(4,4-Dimethyl-2-pentyl)-5,7,7-trimethyl-n-octanoic Acid

Formula: C₁₈H₃₆O₂
Density: 0.88 (20 °C)
Flash Pt: 165 °C
MDL Number: MFCD00135620
CAS Number: 54680-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75efa5e0602cbf75b258de307d6b0ae824a6d52e&vid=75efa5e0602cbf75b258de307d6b0ae824a6d52e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR