Isopropylamine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Propan-2-amine hydrochloride, 2-Aminopropane Hydrochloride
I0166-25G
I0166-500G
I0166-100G
TCI0166-025GEA
53.72
CAD
TCI0166-025G
TCI0166-500G
TCI0166-100G
Isopropylamine hydrochloride ≥98.0%
Isopropylamine hydrochloride
CAS Number: 15572-56-2
MDL Number: MFCD00050705
Molecular Formula: C3H9N
Molecular Weight: 95.57
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 160
Other Product Names / Synonyms: 2-Aminopropane Hydrochloride
Formula:
C₃H₉N Melting Pt: 160 °C |
MDL Number:
MFCD00050705 CAS Number: 15572-56-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...