Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

I0096-25G I0096-500G
TCI0096-025GEA 75.54 CAD
TCI0096-025G TCI0096-500G
Isobutylamine hydrochloride ≥99.0% (by titrimetric analysis)
Isobutylamine hydrochloride

CAS Number: 5041-09-8
MDL Number: MFCD00060199
Molecular Formula: C4H11N
Molecular Weight: 109.60
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 178


Other Product Names / Synonyms: 1-Amino-2-methylpropane Hydrochloride

Formula: C₄H₁₁N
Melting Pt: 178 °C
MDL Number: MFCD00060199
CAS Number: 5041-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74998a3ce37d15f2e38b4f262e2b4e2bbc034d2f&vid=74998a3ce37d15f2e38b4f262e2b4e2bbc034d2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR