β-Ionone ≥95.0%

Supplier: TCI America

I0077-25ML I0077-500ML I0077-100ML
TCI0077-025MLEA 66.36 CAD
TCI0077-025ML TCI0077-500ML TCI0077-100ML
β-Ionone ≥95.0%
β-Ionone

CAS Number: 14901-07-6
MDL Number: MFCD00001549
Molecular Formula: C13H20O
Molecular Weight: 192.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 112
Specific Gravity (20/20): 0.95


Product Categories: Bioscience, Terpenes (Others)
Other Product Names / Synonyms: beta-Jonone

Formula: C₁₃H₂₀O
Boiling Pt: 151 °C
Density: 0.95 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00167092
CAS Number: 14901-07-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6eeb79de71382ac9ac78c2bc6a1ce722259feb3&vid=b6eeb79de71382ac9ac78c2bc6a1ce722259feb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR