Inosine 5'-monophosphate disodium salt hydrate ≥98.0% (by HPLC and UV method)

Supplier: TCI America

I0036-1G I0036-25G I0036-5G
TCI0036-001GEA 35.34 CAD
TCI0036-001G TCI0036-025G TCI0036-5G
Inosine 5'-monophosphate disodium salt hydrate ≥98.0% (by HPLC and UV method)
Inosine 5'-monophosphate disodium salt hydrate

CAS Number: 4691-65-0
MDL Number: MFCD00036201
Molecular Formula: C10H13N4O8P
Molecular Weight: 392.17
Purity/Analysis Method: >98.0% (HPLC,E)
Form: Crystal


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: Disodium Inosinate, Inosic Acid Disodium Salt, 5'-Inosinic Acid Disodium Salt, 5'-IMP 2Na

Formula: C₁₀H₁₃N₄O₈P
MDL Number: MFCD00036201
CAS Number: 4691-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3155c0497ae7766a4c054a039890e6f82561dfb9&vid=3155c0497ae7766a4c054a039890e6f82561dfb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,E)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR