3-Indoleacrylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
IAA, 3-(1-H-Indol-3-yl)-2-propenoic acid, 3-(1H-Indol-3-yl)acrylic acid, 3-(3-Indolyl)acrylic Acid
I0025-1G
I0025-5G
TCI0025-001GEA
106.11
CAD
TCI0025-001G
TCI0025-005G
3-Indoleacrylic acid ≥98.0% (by HPLC, titration analysis)
3-Indoleacrylic acid
CAS Number: 1204-06-4
MDL Number: MFCD00005633
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 3-(3-Indolyl)acrylic Acid, IAA
Formula:
C₁₁H₉NO₂ MW: 187.20 g/mol |
MDL Number:
MFCD00005633 CAS Number: 1204-06-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...