Potassium indole-3-acetate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
K-IAA, 3-Indoleacetic acid potassium salt, Heteroauxine, Indol-3-ylacetic acid potassium salt, Indole-3-acetic acid potassium salt, Potassium 3-Indoleacetate
I0023-25G
I0023-1G
TCI0023-025GEA
292.3
CAD
TCI0023-025G
TCI0023-001G
Potassium indole-3-acetate ≥98.0% (by HPLC, total nitrogen)
Potassium indole-3-acetate
CAS Number: 2338-19-4
MDL Number: MFCD00066213
Molecular Formula: C10H9NO2
Molecular Weight: 213.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: Heteroauxine, 3-Indoleacetic Acid Potassium Salt, K-IAA
Formula:
C₁₀H₈KNO₂ MW: 213.28 g/mol |
MDL Number:
MFCD00066213 CAS Number: 2338-19-4 EINECS: 219-049-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...